Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028766.2 | NP_083042.1 | 6 | 630 | ||
GenBank mRNA | AK004778.1 | - | 6 | 631 | ||
AK037165.1 | - | 6 | 602 | |||
AK050173.1 | - | 6 | 669 | |||
AK151693.1 | - | 6 | 623 | |||
AK151766.1 | - | 6 | 648 | |||
AK152078.1 | - | 6 | 623 | |||
AK168559.1 | - | 6 | 638 | |||
BC005521.1 | - | 6 | 595 | |||
BC024933.1 | - | 6 | 617 | 3990353 |