Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810403A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028814.3 | NP_083090.2 | 3 | 345 | ||
GenBank mRNA | AK012969.1 | - | 3 | 322 | ||
AK083018.1 | - | 3 | 306 | |||
AK170490.1 | - | 3 | 340 | |||
AK173057.1 | - | 3 | 309 | |||
BC006621.1 | - | 3 | 320 | 3158104 | ||
BY097934.1 | - | 3 | 346 | |||
CX562176.1 | - | 3 | 311 |