Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028908.3 | NP_083184.1 | 1 | 2813 | ||
GenBank mRNA | AK042358.1 | - | 1 | 2786 | ||
AK080043.1 | - | 1 | 2767 | |||
AK220388.1 | - | 1 | 2740 |