Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026520.3 | NP_080796.1 | 1 | 1075 | ||
GenBank mRNA | AK010777.1 | - | 1 | 1086 | ||
AK018219.1 | - | 1 | 1026 | |||
AK033646.1 | - | 1 | 1075 | |||
AK077654.1 | - | 1 | 1058 | |||
AK172089.1 | - | 1 | 1060 | |||
BC028637.1 | - | 1 | 1043 | 1248203 |