Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CELF2 SIRNA Assays ›
          Gene Symbol:
          CELF2
          Gene Name:
          CUGBP, Elav-like family member 2
          Gene Aliases:
          BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
          Chromosome Location:
          Chr.10: 10462515 - 11336675 on Build GRCh38
          UniGene ID:
          Hs.309288
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18095

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CELF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025076.2 NP_001020247.1 2, 3 493
          NM_001025077.2 NP_001020248.1 2, 3 350
          NM_001083591.1 NP_001077060.1 2, 3 493
          NM_001326317.1 NP_001313246.1 4, 5 352
          NM_001326318.1 NP_001313247.1 4, 5 352
          NM_001326319.1 NP_001313248.1 5, 6 390
          NM_001326320.1 NP_001313249.1 5, 6 446
          NM_001326321.1 NP_001313250.1 4, 5 352
          NM_001326323.1 NP_001313252.1 5, 6 446
          NM_001326324.1 NP_001313253.1 3, 4 329
          NM_001326325.1 NP_001313254.1 4, 5 423
          NM_001326326.1 NP_001313255.1 3, 4 348
          NM_001326327.1 NP_001313256.1 3, 4 348
          NM_001326328.1 NP_001313257.1 4, 5 442
          NM_001326329.1 NP_001313258.1 2, 3 382
          NM_001326330.1 NP_001313259.1 2, 3 382
          NM_001326331.1 NP_001313260.1 2, 3 534
          NM_001326332.1 NP_001313261.1 2, 3 534
          NM_001326333.1 NP_001313262.1 2, 3 534
          NM_001326334.1 NP_001313263.1 3, 4 719
          NM_001326335.1 NP_001313264.1 2, 3 534
          NM_001326336.1 NP_001313265.1 2, 3 534
          NM_001326337.1 NP_001313266.1 2, 3 534
          NM_001326340.1 NP_001313269.1 2, 3 446
          NM_001326341.1 NP_001313270.1 2, 3 446
          NM_001326342.1 NP_001313271.1 2, 3 446
          NM_001326343.1 NP_001313272.1 2, 3 446
          NM_001326344.1 NP_001313273.1 2, 3 236
          NM_001326345.1 NP_001313274.1 2, 3 314
          NM_001326347.1 NP_001313276.1 2, 3 644
          NM_001326348.1 NP_001313277.1 2, 3 642
          NM_001326349.1 NP_001313278.1 2, 3 642
          NM_006561.3 NP_006552.3 2, 3 439
          XM_005252354.4 XP_005252411.1 2, 3 529
          XM_011519294.2 XP_011517596.1 2, 3 529
          XM_011519297.1 XP_011517599.1 2, 3 652
          XM_017015542.1 XP_016871031.1 1, 2 205
          XM_017015543.1 XP_016871032.1 2, 3 295
          XM_017015544.1 XP_016871033.1 2, 3 295
          XM_017015545.1 XP_016871034.1 2, 3 295
          XM_017015546.1 XP_016871035.1 2, 3 295
          XM_017015547.1 XP_016871036.1 2, 3 529
          XM_017015548.1 XP_016871037.1 2, 3 295
          XM_017015549.1 XP_016871038.1 2, 3 295
          XM_017015550.1 XP_016871039.1 2, 3 1275
          XM_017015551.1 XP_016871040.1 2, 3 529
          XM_017015552.1 XP_016871041.1 2, 3 652
          XM_017015553.1 XP_016871042.1 2, 3 652
          XM_017015554.1 XP_016871043.1 2, 3 652
          XM_017015555.1 XP_016871044.1 1, 2 205
          XM_017015564.1 XP_016871053.1 1, 2 225
          XM_017015566.1 XP_016871055.1 1, 2 225
          XM_017015567.1 XP_016871056.1 2, 3 365
          XM_017015568.1 XP_016871057.1 4, 5 352
          XM_017015569.1 XP_016871058.1 2, 3 652
          XM_017015571.1 XP_016871060.1 2, 3 652
          XM_017015572.1 XP_016871061.1 2, 3 353
          GenBank mRNA AF036956.1 - 2, 3 409
          AF090693.1 - 2, 3 325
          AF090694.1 - 2, 3 277
          AF432906.1 - 2, 3 345
          AK293164.1 - 2, 3 386
          AK299548.1 - 2, 3 269
          AK299983.1 - 2, 3 558
          AK299992.1 - 2, 3 426
          AK304216.1 - 1, 2 235
          AK314087.1 - 2, 3 448
          AK316354.1 - 2, 3 245
          BC036391.1 - 2, 3 411 5260623
          BP229421.1 - 2, 3 350
          BQ213648.1 - 2, 3 494
          DA819821.1 - 1, 2 163
          DB478862.1 - 2, 3 439
          U69546.1 - 2, 3 226

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017197, NM_001083586, XM_008771864, XM_008771861, XM_008771863, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001110231, NM_001110228, NM_001110230, NM_001160292, NM_001110229, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          mRNA processing
          regulation of heart contraction
          nucleotide binding
          RNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0