Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007225.1 | NP_001007226.1 | 15 | 2980 | ||
NM_001291869.1 | NP_001278798.1 | 16 | 3127 | |||
NM_001291872.1 | NP_001278801.1 | 15 | 2929 | |||
NM_001291873.1 | NP_001278802.1 | 15 | 2911 | |||
NM_001291874.1 | NP_001278803.1 | 14 | 2782 | |||
NM_001291875.1 | NP_001278804.1 | 14 | 2847 | |||
NM_006548.4 | NP_006539.3 | 16 | 3109 | |||
XM_006713477.3 | XP_006713540.1 | 11 | 2533 | |||
XM_006713478.3 | XP_006713541.1 | 10 | 2399 | |||
XM_017005557.1 | XP_016861046.1 | 16 | 3106 | |||
XM_017005558.1 | XP_016861047.1 | 15 | 3003 | |||
XM_017005559.1 | XP_016861048.1 | 15 | 3001 | |||
XM_017005560.1 | XP_016861049.1 | 16 | 3029 | |||
GenBank mRNA | BC021290.2 | - | 16 | 3093 | 5090334 | |
BX537583.1 | - | 1 | 2318 |