Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267608.1 | NP_001254537.1 | 9 | 2080 | ||
NM_006589.2 | NP_006580.2 | 10 | 2134 | |||
NM_198264.1 | NP_937995.1 | 7 | 1846 | |||
XM_005244845.1 | XP_005244902.1 | 10 | 2131 | |||
GenBank mRNA | AF070550.1 | - | 4 | 529 | ||
AL353941.1 | - | 5 | 918 | |||
BC006297.1 | - | 2 | 546 | |||
BC006493.2 | - | 10 | 1614 | 2966515 | ||
BC007444.2 | - | 10 | 1614 | 2966515 | ||
BC010471.1 | - | 10 | 2038 | 4179576 | ||
BC017262.2 | - | 3 | 665 | |||
BC040136.1 | - | 7 | 1323 | 6061151 |