Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025607.3 | NP_079883.3 | 2 | 242 | ||
GenBank mRNA | AK010854.1 | - | 2 | 255 | ||
AK018088.1 | - | 1 | 687 | |||
AK018849.1 | - | 3 | 363 | |||
AK159788.1 | - | 2 | 255 | |||
AK166904.1 | - | 2 | 254 | |||
AK166995.1 | - | 2 | 255 | |||
AK167396.1 | - | 2 | 255 | |||
AK167633.1 | - | 2 | 255 | |||
AK167935.1 | - | 2 | 255 | |||
AK168670.1 | - | 2 | 256 | |||
BC005559.1 | - | 2 | 266 | |||
BC083119.1 | - | 2 | 243 | 5374213 |