Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025998.3 | NP_080274.2 | 7 | 1096 | ||
GenBank mRNA | AK011850.1 | - | 7 | 1027 | ||
AK076029.1 | - | 4 | 578 | |||
EF058048.2 | - | 7 | 1149 |