Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF266 SIRNA Assays ›
Gene Symbol:
ZNF266
Gene Name:
zinc finger protein 266
Gene Aliases:
HZF1
Chromosome Location:
Chr.19: 9412426 - 9435578 on Build GRCh38
UniGene ID:
Hs.656185
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18407
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF266
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271314.1
NP_001258243.1
11
3005
NM_006631.3
NP_006622.2
11
3056
XM_005259716.3
XP_005259773.1
8
2722
XM_006722620.2
XP_006722683.1
11
2788
XM_006722621.2
XP_006722684.1
10
2694
XM_006722622.2
XP_006722685.1
9
2533
XM_006722623.3
XP_006722686.1
8
2468
XM_006722627.2
XP_006722690.1
10
3385
XM_006722630.2
XP_006722693.1
9
2699
XM_011527644.1
XP_011525946.1
10
2704
XM_011527647.1
XP_011525949.1
11
3243
XM_011527648.1
XP_011525950.1
11
3239
XM_011527650.1
XP_011525952.1
11
3042
XM_017026154.1
XP_016881643.1
11
2813
XM_017026155.1
XP_016881644.1
11
2810
XM_017026156.1
XP_016881645.1
12
3113
XM_017026157.1
XP_016881646.1
10
3388
XM_017026158.1
XP_016881647.1
12
3084
XM_017026159.1
XP_016881648.1
12
3106
XM_017026160.1
XP_016881649.1
9
3279
XM_017026161.1
XP_016881650.1
9
2984
XM_017026162.1
XP_016881651.1
10
2860
XM_017026163.1
XP_016881652.1
10
2829
XM_017026164.1
XP_016881653.1
8
2634
XM_017026165.1
XP_016881654.1
11
2803
XM_017026166.1
XP_016881655.1
8
2624
XM_017026167.1
XP_016881656.1
8
2458
XM_017026168.1
XP_016881657.1
9
3284
XM_017026169.1
XP_016881658.1
7
2830
XM_017026170.1
XP_016881659.1
8
2444
XM_017026171.1
XP_016881660.1
7
2379
XM_017026172.1
XP_016881661.1
7
2369
XM_017026173.1
XP_016881662.1
10
2724
GenBank mRNA
AB095928.1
-
5
3841
AK027480.1
-
1
1294
AL833471.1
-
4
1961
AL833503.1
-
11
2735
AY376240.1
-
10
2761
BC017407.1
-
1
999
BC071628.1
-
1
1312
BC112919.1
-
9
2945
7502631
BC126255.1
-
4
2031
8991971
BC143940.1
-
4
2031
BC146837.1
-
5
1901
9056764
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix