Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLIC4 SIRNA Assays ›
          Gene Symbol:
          Clic4
          Gene Name:
          chloride intracellular channel 4 (mitochondrial)
          Gene Aliases:
          D0Jmb3, TU-74, mc3s5, mtCLIC
          Chromosome Location:
          Chr.4: 135213969 - 135272760 on Build GRCm38
          UniGene ID:
          Mm.257765
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 184138

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Clic4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013885.2 NP_038913.1 2, 3 346
          GenBank mRNA AF102578.1 - 2, 3 344
          AK031211.1 - 2, 3 353
          AK049191.1 - 2, 3 351
          AK049255.1 - 2, 3 351
          AK054024.1 - 2, 3 351
          AK150993.1 - 2, 3 350
          AK152273.1 - 2, 3 352
          AK153432.1 - 2, 3 353
          AK155826.1 - 2, 3 350
          AK159402.1 - 2, 3 353
          AK160856.1 - 2, 3 352
          AK161034.1 - 2, 3 352
          AK166244.1 - 2, 3 353
          AK166794.1 - 2, 3 350
          AK169376.1 - 2, 3 350
          AK169834.1 - 2, 3 350
          AK170942.1 - 2, 3 401
          AK171243.1 - 2, 3 353
          AK172026.1 - 2, 3 352
          AK172516.1 - 2, 3 326
          AK172660.1 - 2, 3 350
          AK186780.1 - 170
          AK211909.1 - 1, 2 53
          BC046384.1 - 2, 3 330 3155573
          BC052890.1 - 2, 3 334 30056606
          CT010279.1 - 2, 3 175

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          endothelial cell morphogenesis
          transport
          ion transport
          chloride transport
          apoptotic process
          vacuolar acidification
          fertilization
          keratinocyte differentiation
          negative regulation of cell migration
          regulation of ion transmembrane transport
          multicellular organism growth
          branching morphogenesis of an epithelial tube
          retina vasculature morphogenesis in camera-type eye
          cellular response to calcium ion
          chloride transmembrane transport
          voltage-gated ion channel activity
          chloride channel activity
          protein binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0