Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017382.5 | NP_059078.2 | 5 | 716 | ||
GenBank mRNA | AB232605.1 | - | 5 | 583 | ||
AK014268.1 | - | 5 | 717 | |||
AK031143.1 | - | 5 | 619 | |||
AK132072.1 | - | 5 | 700 | |||
AK132159.1 | - | 5 | 702 | |||
AK147122.1 | - | 5 | 717 | |||
BC010722.1 | - | 5 | 705 | 2811973 | ||
D50500.1 | - | 3 | 224 |