Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020003.1 | NP_064387.1 | 3 | 543 | ||
GenBank mRNA | AB027141.1 | - | 3 | 499 | ||
AB027142.1 | - | 3 | 543 | |||
AK002728.1 | - | 3 | 507 | |||
AK007803.1 | - | 3 | 516 | |||
AK075614.1 | - | 3 | 521 | |||
AK080538.1 | - | 3 | 517 | |||
AK150794.1 | - | 3 | 516 | |||
AK154340.1 | - | 3 | 516 | |||
AK160793.1 | - | 483 | ||||
AK170696.1 | - | 3 | 506 | |||
AK171042.1 | - | 3 | 515 | |||
AK171142.1 | - | 1172 | ||||
AK172606.1 | - | 1 | 1165 | |||
AK182742.1 | - | 2 | 349 | |||
BC021547.1 | - | 3 | 509 | 4507218 |