Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020497.2 | NP_065243.2 | 11 | 2417 | ||
GenBank mRNA | AF178935.1 | - | 10 | 1459 | ||
AK028939.1 | - | 11 | 2370 | |||
AK083110.1 | - | 2549 | ||||
AK133363.1 | - | 11 | 2418 | |||
AK163486.1 | - | 2 | 408 | |||
BC066056.1 | - | 11 | 2402 | |||
BC078415.1 | - | 10 | 1871 | |||
BC138579.1 | - | 10 | 1409 | 8861599 | ||
BC138580.1 | - | 10 | 1409 | 8861600 | ||
BC145339.1 | - | 9 | 1445 | |||
BC145340.1 | - | 9 | 1479 |