Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009441.2 | NP_033467.2 | 18 | 1614 | ||
GenBank mRNA | AB008516.1 | - | 17 | 1491 | ||
AK053765.1 | - | 18 | 1718 | |||
AK084201.1 | - | 8 | 649 | |||
AK088273.1 | - | 17 | 1561 | |||
AK165850.1 | - | 7 | 522 | |||
AK166120.1 | - | 8 | 554 | |||
AK166164.1 | - | 9 | 703 | |||
AK220495.1 | - | 18 | 1656 | |||
BC019173.1 | - | 18 | 1583 | |||
BC037654.1 | - | 17 | 1516 | |||
BC057207.1 | - | 17 | 1518 | |||
BC145489.1 | - | 13 | 1134 |