Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005368.2 | NP_001005368.1 | 3 | 410 | ||
NM_001324164.1 | NP_001311093.1 | 3 | 378 | |||
NM_001324165.1 | NP_001311094.1 | 3 | 516 | |||
NM_001324166.1 | NP_001311095.1 | 3 | 280 | |||
NM_001324167.1 | NP_001311096.1 | 3 | 276 | |||
NM_001324168.1 | NP_001311097.1 | 3 | 414 | |||
NM_006973.2 | NP_008904.1 | 3 | 272 | |||
XM_017016616.1 | XP_016872105.1 | 2 | 3343 | |||
GenBank mRNA | AI871265.1 | - | 3 | 315 | ||
AK312132.1 | - | 3 | 269 | |||
BC022842.1 | - | 2 | 207 | 5014207 | ||
BG477527.1 | - | 3 | 274 | |||
HY045180.1 | - | 3 | 415 | |||
U69645.1 | - | 3 | 248 |