Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPARGC1A SIRNA Assays ›
          Gene Symbol:
          Ppargc1a
          Gene Name:
          peroxisome proliferator-activated receptor gamma, coactivator 1 alpha
          Gene Aliases:
          Ppargc1
          Chromosome Location:
          Chr.14: 63095291 - 63190688 on Build Rnor_6.0
          UniGene ID:
          Rn.19172
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 189917

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ppargc1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031347.1 NP_112637.1 5, 6 800
          XM_008770219.1 XP_008768441.1 5, 6 867
          XM_008770220.1 XP_008768442.1 4, 5 2037
          GenBank mRNA AB025784.1 - 5, 6 800
          AY237127.1 - 5, 6 759

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_013261, AF159714, JQ772117, AF186379, AK296591, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008904, AF049330, AB061324, AK138668, AK177938, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          response to dietary excess
          galactose metabolic process
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          mitochondrion organization
          androgen metabolic process
          response to cold
          positive regulation of mitochondrion organization
          response to organic cyclic compound
          response to activity
          response to muscle activity
          negative regulation of smooth muscle cell migration
          respiratory electron transport chain
          positive regulation of cellular metabolic process
          response to nutrient levels
          circadian regulation of gene expression
          cellular response to oxidative stress
          response to statin
          response to drug
          response to starvation
          regulation of circadian rhythm
          response to leucine
          negative regulation of neuron apoptotic process
          cellular respiration
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          positive regulation of smooth muscle cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          flavone metabolic process
          cellular response to nitrite
          cellular response to tumor necrosis factor
          cellular response to fatty acid
          cellular response to hypoxia
          response to epinephrine
          response to norepinephrine
          cellular response to thyroid hormone stimulus
          negative regulation of neuron death
          positive regulation of cellular respiration
          positive regulation of mitochondrial DNA metabolic process
          positive regulation of muscle tissue development
          negative regulation of receptor activity
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of energy homeostasis
          positive regulation of ATP biosynthetic process
          nucleotide binding
          RNA polymerase II transcription cofactor activity
          DNA binding
          chromatin binding
          transcription coactivator activity
          RNA binding
          protein binding
          transcription factor binding
          ligand-dependent nuclear receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          chromatin DNA binding
          ubiquitin protein ligase binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0