Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19080
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
1
1112
NM_001258250.1
NP_001245179.1
1
1112
NM_001258251.1
NP_001245180.1
1
1112
NM_001258252.1
NP_001245181.1
1
1112
NM_001258253.1
NP_001245182.1
1
1112
NM_001258254.1
NP_001245183.1
1
1112
NM_001258255.1
NP_001245184.1
1
1112
NM_001258256.1
NP_001245185.1
1
1112
NM_001258257.1
NP_001245186.1
1
1112
NM_001258258.1
NP_001245187.1
1
1112
NM_001258259.1
NP_001245188.1
1
1112
NM_001258260.1
NP_001245189.1
1
1112
NM_001258261.1
NP_001245190.1
1
1112
NM_001258262.1
NP_001245191.1
1
1112
NM_001258263.1
NP_001245192.1
1
1112
NM_001258264.1
NP_001245193.1
1
1112
NM_001258265.1
NP_001245194.1
1
1112
NM_001258266.1
NP_001245195.1
1
1112
NM_001258267.1
NP_001245196.1
1
1112
NM_001258268.1
NP_001245197.1
1
1112
NM_001258269.1
NP_001245198.1
1
1112
NM_001258270.1
NP_001245199.1
1
1112
NM_007125.4
NP_009056.3
1
1112
NM_182659.1
NP_872600.1
1
1112
NM_182660.1
NP_872601.1
1
1112
NR_047596.1
1
1112
NR_047597.1
1
1112
NR_047598.1
1
1112
NR_047599.1
1
1112
NR_047600.1
1
1112
NR_047601.1
1
1112
NR_047602.1
1
1112
NR_047603.1
1
1112
NR_047604.1
1
1112
NR_047605.1
1
1112
NR_047606.1
1
1112
NR_047607.1
1
1112
NR_047608.1
1
1112
NR_047609.1
1
1112
NR_047610.1
1
1112
NR_047611.1
1
1112
NR_047612.1
1
1112
NR_047613.1
1
1112
NR_047614.1
1
1112
NR_047615.1
1
1112
NR_047616.1
1
1112
NR_047617.1
1
1112
NR_047618.1
1
1112
NR_047619.1
1
1112
NR_047620.1
1
1112
NR_047621.1
1
1112
NR_047622.1
1
1112
NR_047623.1
1
1112
NR_047624.1
1
1112
NR_047625.1
1
1112
NR_047626.1
1
1112
NR_047627.1
1
1112
NR_047628.1
1
1112
NR_047629.1
1
1112
NR_047630.1
1
1112
NR_047631.1
1
1112
NR_047632.1
1
1112
NR_047633.1
1
1112
NR_047634.1
1
1112
NR_047635.1
1
1112
NR_047636.1
1
1112
NR_047637.1
1
1112
NR_047638.1
1
1112
NR_047639.1
1
1112
NR_047640.1
1
1112
NR_047641.1
1
1112
NR_047642.1
1
1112
NR_047643.1
1
1112
NR_047644.1
1
1112
NR_047645.1
1
1112
NR_047646.1
1
1112
NR_047647.1
1
1112
XM_005262518.3
XP_005262575.1
1
458
XM_006724875.3
XP_006724938.1
1
458
XM_011531441.2
XP_011529743.1
1
458
XM_011531442.2
XP_011529744.1
1
458
XM_011531443.2
XP_011529745.1
1
458
XM_011531445.2
XP_011529747.1
1
458
XM_011531446.2
XP_011529748.1
1
458
XM_011531447.2
XP_011529749.1
1
458
XM_011531448.2
XP_011529750.1
1
458
XM_011531449.2
XP_011529751.1
1
458
XM_011531450.2
XP_011529752.1
1
458
XM_011531451.2
XP_011529753.1
1
458
XM_011531452.2
XP_011529754.1
1
458
XM_011531453.2
XP_011529755.1
1
458
XM_011531454.2
XP_011529756.1
1
458
XM_011531455.2
XP_011529757.1
1
458
XM_011531457.2
XP_011529759.1
1
458
XM_011531458.2
XP_011529760.1
1
458
XM_011531459.2
XP_011529761.1
1
458
XM_011531460.2
XP_011529762.1
1
458
XM_011531461.2
XP_011529763.1
1
458
XM_011531462.2
XP_011529764.1
1
458
XM_011531463.2
XP_011529765.1
1
458
XM_011531464.2
XP_011529766.1
1
458
XM_011531465.2
XP_011529767.1
1
458
XM_017030066.1
XP_016885555.1
1
458
XM_017030067.1
XP_016885556.1
1
458
XM_017030068.1
XP_016885557.1
1
458
XM_017030069.1
XP_016885558.1
1
458
XM_017030070.1
XP_016885559.1
1
458
XM_017030071.1
XP_016885560.1
1
458
XM_017030072.1
XP_016885561.1
1
458
XM_017030073.1
XP_016885562.1
1
458
XM_017030074.1
XP_016885563.1
1
458
GenBank mRNA
AF000994.1
-
1
1112
AF000995.1
-
1
1112
AF000996.1
-
1
1112
AK292858.1
-
1
362
AY682463.1
-
1
113
AY730613.1
-
1
107
AY769061.1
-
1
107
AY769062.1
-
1
107
AY769063.1
-
1
107
AY769064.1
-
1
107
AY769065.1
-
1
107
AY769066.1
-
1
107
AY769067.1
-
1
107
AY769068.1
-
1
107
AY769069.1
-
1
107
AY934848.1
-
1
107
AY934849.1
-
1
107
AY934850.1
-
1
107
AY934851.1
-
1
107
AY934852.1
-
1
107
AY934853.1
-
1
107
AY934854.1
-
1
107
AY934855.1
-
1
107
AY934856.1
-
1
107
AY934857.1
-
1
107
BC071744.1
-
1
364
DQ012094.1
-
1
107
DQ140388.1
-
1
107
DQ140389.1
-
1
107
DQ140390.1
-
1
107
DQ341385.1
-
1
107
DQ341386.1
-
1
107
DQ341387.1
-
1
107
DQ345542.1
-
1
107
DQ345543.1
-
1
107
DQ345544.1
-
1
107
DQ350456.1
-
1
107
DQ350457.1
-
1
107
DQ350458.1
-
1
107
DQ350459.1
-
1
107
EF612660.1
-
1
107
EF612661.1
-
1
107
EF612662.1
-
1
107
EF612663.1
-
1
107
EF612664.1
-
1
107
EF612665.1
-
1
107
EF612666.1
-
1
107
EF612667.1
-
1
107
EF612668.1
-
1
107
EF612669.1
-
1
107
EF612670.1
-
1
107
EF612671.1
-
1
107
EF612672.1
-
1
107
EF612673.1
-
1
107
EF612674.1
-
1
107
EF612675.1
-
1
107
EF612677.1
-
1
107
EF612678.1
-
1
107
EF612679.1
-
1
107
EF612680.1
-
1
107
EF612681.1
-
1
107
EF612682.1
-
1
107
EF612683.1
-
1
107
EF612684.1
-
1
107
EF612685.1
-
1
107
EF612686.1
-
1
107
EF612687.1
-
1
107
EF612688.1
-
1
107
EF612689.1
-
1
107
EF612690.1
-
1
107
EF612691.1
-
1
107
EF612692.1
-
1
107
EF612693.1
-
1
107
EF612694.1
-
1
107
EF612695.1
-
1
107
EF612696.1
-
1
107
EF612697.1
-
1
107
EF612698.1
-
1
107
EF612699.1
-
1
107
EF653165.1
-
1
107
EF653166.1
-
1
107
EF653167.1
-
1
107
EF653168.1
-
1
107
EF653169.1
-
1
107
EF653170.1
-
1
107
EF653171.1
-
1
107
EF653172.1
-
1
107
EF653173.1
-
1
107
EF653174.1
-
1
107
EF653175.1
-
1
107
EF653176.1
-
1
107
EF653177.1
-
1
107
EF653178.1
-
1
107
EF653179.1
-
1
107
EF653180.1
-
1
107
EF653181.1
-
1
107
EF653182.1
-
1
107
EF653183.1
-
1
107
EF653185.1
-
1
107
EF653186.1
-
1
107
EF653187.1
-
1
107
EF653188.1
-
1
107
EF653189.1
-
1
107
EF653190.1
-
1
107
EF653191.1
-
1
107
EF653192.1
-
1
107
EF653193.1
-
1
107
EF653194.1
-
1
107
EF653195.1
-
1
107
EF653197.1
-
1
107
EF653198.1
-
1
107
EF653199.1
-
1
107
EF653200.1
-
1
107
EF653201.1
-
1
107
EF653202.1
-
1
107
EF653203.1
-
1
107
EF653204.1
-
1
107
EF653205.1
-
1
107
EF653206.1
-
1
107
EF653207.1
-
1
107
EF653208.1
-
1
107
EF653209.1
-
1
107
EF653210.1
-
1
107
EF653211.1
-
1
107
EF653212.1
-
1
107
EF653213.1
-
1
107
EF653214.1
-
1
107
EF653215.1
-
1
107
EF653216.1
-
1
107
EF653217.1
-
1
107
EF653218.1
-
1
107
EF653219.1
-
1
107
EF653220.1
-
1
107
EF653221.1
-
1
107
EF653223.1
-
1
107
EF653224.1
-
1
107
EF685283.1
-
1
107
EF685284.1
-
1
107
EF685285.1
-
1
107
EF685286.1
-
1
107
EF685287.1
-
1
107
EF685288.1
-
1
107
EF685289.1
-
1
107
EF685290.1
-
1
107
EF685291.1
-
1
107
EF685292.1
-
1
107
EF685294.1
-
1
107
EF685295.1
-
1
107
EF685296.1
-
1
107
EF685297.1
-
1
107
EF685298.1
-
1
107
EF685299.1
-
1
107
EF685300.1
-
1
107
EF685301.1
-
1
107
EF685302.1
-
1
107
EF685303.1
-
1
107
EF685304.1
-
1
107
EF685305.1
-
1
107
EF685306.1
-
1
107
EF685307.1
-
1
107
EF685309.1
-
1
107
EF685310.1
-
1
107
EF685311.1
-
1
107
EF685312.1
-
1
107
EF685313.1
-
1
107
EF685314.1
-
1
107
EF685315.1
-
1
107
EF685316.1
-
1
107
EF685317.1
-
1
107
EF685318.1
-
1
107
EF685319.1
-
1
107
EF685320.1
-
1
107
EF685321.1
-
1
107
EF685322.1
-
1
107
EF685323.1
-
1
107
EF685324.1
-
1
107
EF685326.1
-
1
107
EF685327.1
-
1
107
EF685328.1
-
1
107
EF685329.1
-
1
107
EF685330.1
-
1
107
EF685331.1
-
107
EF685332.1
-
1
107
EF685333.1
-
1
107
EF685334.1
-
1
107
EF685335.1
-
1
107
EF685336.1
-
1
107
EF685337.1
-
1
107
EF685338.1
-
1
107
EF685339.1
-
1
107
EF685340.1
-
1
107
EF685341.1
-
1
107
EF685342.1
-
1
107
EU026275.1
-
1
107
EU026276.1
-
1
107
EU026277.1
-
1
107
EU026278.1
-
1
107
EU026279.1
-
1
107
EU026281.1
-
1
107
EU026282.1
-
1
107
EU026284.1
-
1
107
EU026285.1
-
1
107
EU026286.1
-
1
107
EU026287.1
-
1
107
EU026288.1
-
1
107
EU026289.1
-
1
107
EU026290.1
-
1
107
EU026292.1
-
1
107
EU026293.1
-
1
107
EU026294.1
-
1
107
EU026295.1
-
1
107
EU026296.1
-
1
107
EU026297.1
-
1
107
EU026298.1
-
1
107
EU026299.1
-
1
107
EU026300.1
-
1
107
EU026301.1
-
1
107
EU026302.1
-
1
107
EU026303.1
-
1
107
EU026305.1
-
1
107
EU026306.1
-
1
107
EU026307.1
-
1
107
EU026308.1
-
1
107
EU026309.1
-
1
107
EU026310.1
-
1
107
EU026311.1
-
1
107
EU026312.1
-
1
107
EU026313.1
-
1
107
EU026315.1
-
1
107
EU026316.1
-
1
107
EU026317.1
-
1
107
EU026318.1
-
1
107
EU026319.1
-
1
107
EU026320.1
-
1
107
EU026321.1
-
1
107
EU026322.1
-
1
107
EU026323.1
-
1
107
EU026324.1
-
1
107
EU026325.1
-
1
107
EU026326.1
-
1
107
EU026327.1
-
1
107
EU026328.1
-
1
107
EU026329.1
-
1
107
EU026330.1
-
1
107
EU026331.1
-
1
107
EU026332.1
-
1
107
EU026333.1
-
1
107
EU026334.1
-
1
107
EU026335.1
-
1
107
EU026336.1
-
1
107
EU026337.1
-
1
107
EU026338.1
-
1
107
EU026339.1
-
1
107
EU026340.1
-
1
107
EU026341.1
-
1
107
EU026342.1
-
1
107
EU026343.1
-
1
107
EU026344.1
-
1
107
EU026345.1
-
1
107
EU026346.1
-
1
107
EU026347.1
-
1
107
EU026348.1
-
1
107
EU026349.1
-
1
107
EU026350.1
-
1
107
EU026351.1
-
1
107
EU116345.1
-
1
107
EU116346.1
-
1
107
EU116347.1
-
1
107
EU116349.1
-
1
107
EU116352.1
-
1
107
EU116353.1
-
1
107
EU163941.1
-
1
107
EU163942.1
-
1
107
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix