Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181627.2 | NP_853658.1 | 396 | |||
GenBank mRNA | AY291062.1 | - | 396 | |||
AY291063.2 | - | 591 | ||||
AY291064.2 | - | 408 | ||||
AY291065.2 | - | 757 | ||||
AY919342.1 | - | 213 | ||||
AY919343.1 | - | 375 | ||||
FJ517605.1 | - | 268 | ||||
FJ517606.1 | - | 110 | ||||
FJ517607.1 | - | 2 | 536 | |||
FQ213378.1 | - | 413 | ||||
FQ213584.1 | - | 413 | ||||
FQ214037.1 | - | 414 | ||||
FQ214498.1 | - | 424 | ||||
FQ229190.1 | - | 413 | ||||
FQ232927.1 | - | 413 |