Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19173
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
9
1679
NM_001258250.1
NP_001245179.1
9
1679
NM_001258251.1
NP_001245180.1
9
1679
NM_001258252.1
NP_001245181.1
9
1679
NM_001258253.1
NP_001245182.1
9
1679
NM_001258254.1
NP_001245183.1
9
1679
NM_001258255.1
NP_001245184.1
9
1679
NM_001258256.1
NP_001245185.1
9
1679
NM_001258257.1
NP_001245186.1
9
1679
NM_001258258.1
NP_001245187.1
9
1679
NM_001258259.1
NP_001245188.1
9
1679
NM_001258260.1
NP_001245189.1
9
1679
NM_001258261.1
NP_001245190.1
9
1679
NM_001258262.1
NP_001245191.1
9
1679
NM_001258263.1
NP_001245192.1
9
1679
NM_001258264.1
NP_001245193.1
9
1679
NM_001258265.1
NP_001245194.1
5
1430
NM_001258266.1
NP_001245195.1
9
1679
NM_001258267.1
NP_001245196.1
9
1679
NM_001258268.1
NP_001245197.1
9
1679
NM_001258269.1
NP_001245198.1
9
1679
NM_001258270.1
NP_001245199.1
9
1679
NM_007125.4
NP_009056.3
9
1679
NM_182659.1
NP_872600.1
9
1679
NM_182660.1
NP_872601.1
9
1679
NR_047596.1
9
1679
NR_047597.1
10
1785
NR_047598.1
10
1785
NR_047599.1
9
1679
NR_047600.1
8
1644
NR_047601.1
9
1679
NR_047602.1
8
1644
NR_047603.1
10
1802
NR_047604.1
9
1679
NR_047605.1
9
1679
NR_047606.1
9
1679
NR_047607.1
9
1679
NR_047608.1
9
1679
NR_047609.1
9
1679
NR_047610.1
9
1679
NR_047611.1
9
1679
NR_047612.1
9
1679
NR_047613.1
8
1629
NR_047614.1
10
1793
NR_047615.1
9
1679
NR_047616.1
10
1793
NR_047617.1
10
1785
NR_047618.1
9
1679
NR_047619.1
9
1679
NR_047620.1
9
1679
NR_047621.1
9
1750
NR_047622.1
9
1679
NR_047623.1
9
1679
NR_047624.1
11
1899
NR_047625.1
9
1679
NR_047626.1
9
1679
NR_047627.1
8
1644
NR_047628.1
9
1679
NR_047629.1
9
1679
NR_047630.1
9
1679
NR_047631.1
9
1679
NR_047632.1
10
1759
NR_047633.1
9
1679
NR_047634.1
8
1644
NR_047635.1
9
1679
NR_047636.1
9
1679
NR_047637.1
9
1679
NR_047638.1
9
1750
NR_047640.1
10
1785
NR_047641.1
9
1679
NR_047642.1
9
1679
NR_047643.1
9
1679
NR_047644.1
9
1679
NR_047645.1
9
1750
NR_047646.1
9
1679
NR_047647.1
9
1679
XM_005262518.3
XP_005262575.1
9
1025
XM_006724875.3
XP_006724938.1
9
1025
XM_011531441.2
XP_011529743.1
9
1025
XM_011531442.2
XP_011529744.1
9
1025
XM_011531443.2
XP_011529745.1
9
1025
XM_011531445.2
XP_011529747.1
9
1025
XM_011531446.2
XP_011529748.1
9
1025
XM_011531447.2
XP_011529749.1
9
1025
XM_011531448.2
XP_011529750.1
9
1025
XM_011531449.2
XP_011529751.1
9
1025
XM_011531450.2
XP_011529752.1
9
1025
XM_011531451.2
XP_011529753.1
9
1025
XM_011531452.2
XP_011529754.1
9
1025
XM_011531453.2
XP_011529755.1
9
1025
XM_011531454.2
XP_011529756.1
9
1025
XM_011531455.2
XP_011529757.1
9
1025
XM_011531457.2
XP_011529759.1
9
1025
XM_011531458.2
XP_011529760.1
9
1025
XM_011531459.2
XP_011529761.1
9
1025
XM_011531460.2
XP_011529762.1
9
1025
XM_011531461.2
XP_011529763.1
9
1025
XM_011531462.2
XP_011529764.1
9
1025
XM_011531463.2
XP_011529765.1
9
1025
XM_011531464.2
XP_011529766.1
9
1025
XM_011531465.2
XP_011529767.1
9
1025
XM_017030066.1
XP_016885555.1
9
1025
XM_017030067.1
XP_016885556.1
9
1025
XM_017030068.1
XP_016885557.1
9
1025
XM_017030069.1
XP_016885558.1
9
1025
XM_017030070.1
XP_016885559.1
9
1025
XM_017030071.1
XP_016885560.1
9
1025
XM_017030072.1
XP_016885561.1
9
1025
XM_017030073.1
XP_016885562.1
9
1025
XM_017030074.1
XP_016885563.1
9
1025
GenBank mRNA
AF000994.1
-
9
1679
AF000995.1
-
9
1679
AF000996.1
-
9
1679
AK292858.1
-
9
929
AY682463.1
-
9
680
AY730613.1
-
9
674
AY769061.1
-
10
780
AY769062.1
-
10
780
AY769063.1
-
8
615
AY769064.1
-
8
639
AY769065.1
-
9
674
AY769066.1
-
9
674
AY769067.1
-
9
674
AY769069.1
-
8
639
AY934848.1
-
9
674
AY934849.1
-
10
780
AY934850.1
-
9
674
AY934851.1
-
9
674
AY934853.1
-
10
754
AY934854.1
-
9
674
AY934855.1
-
9
674
AY934856.1
-
8
639
AY934857.1
-
10
788
DQ140388.1
-
9
674
DQ140389.1
-
9
674
DQ140390.1
-
9
674
DQ341386.1
-
9
674
DQ341387.1
-
9
674
DQ345542.1
-
8
639
DQ345543.1
-
9
674
DQ350457.1
-
9
674
DQ350458.1
-
10
788
DQ350459.1
-
9
674
EF612660.1
-
9
674
EF612661.1
-
9
674
EF612662.1
-
9
674
EF612663.1
-
9
674
EF612664.1
-
9
674
EF612665.1
-
9
674
EF612666.1
-
7
584
EF612667.1
-
9
674
EF612668.1
-
9
674
EF612669.1
-
9
674
EF612670.1
-
9
674
EF612671.1
-
9
674
EF612672.1
-
9
674
EF612673.1
-
9
674
EF612674.1
-
9
674
EF612675.1
-
9
674
EF612676.1
-
9
674
EF612677.1
-
9
674
EF612678.1
-
9
674
EF612679.1
-
9
674
EF612680.1
-
9
674
EF612683.1
-
9
674
EF612684.1
-
9
674
EF612685.1
-
9
674
EF612686.1
-
9
674
EF612687.1
-
9
674
EF612688.1
-
9
674
EF612689.1
-
9
674
EF612690.1
-
9
674
EF612691.1
-
9
674
EF612692.1
-
7
584
EF612693.1
-
9
674
EF612694.1
-
9
674
EF612695.1
-
9
674
EF612696.1
-
9
674
EF612697.1
-
9
674
EF612698.1
-
7
584
EF612699.1
-
9
674
EF653165.1
-
8
639
EF653167.1
-
8
639
EF653168.1
-
9
674
EF653169.1
-
8
639
EF653170.1
-
8
639
EF653171.1
-
9
674
EF653172.1
-
8
565
EF653173.1
-
9
674
EF653174.1
-
10
780
EF653175.1
-
10
780
EF653176.1
-
8
624
EF653177.1
-
9
674
EF653178.1
-
12
1031
EF653179.1
-
9
674
EF653180.1
-
10
780
EF653181.1
-
9
674
EF653182.1
-
8
639
EF653183.1
-
10
780
EF653184.1
-
10
788
EF653186.1
-
10
797
EF653187.1
-
9
674
EF653188.1
-
10
780
EF653189.1
-
5
425
EF653190.1
-
9
674
EF653194.1
-
10
754
EF653195.1
-
10
780
EF653196.1
-
9
674
EF653197.1
-
8
639
EF653198.1
-
9
674
EF653199.1
-
11
894
EF653200.1
-
9
674
EF653201.1
-
8
639
EF653203.1
-
8
639
EF653204.1
-
10
790
EF653205.1
-
9
674
EF653206.1
-
9
674
EF653207.1
-
9
674
EF653208.1
-
10
780
EF653209.1
-
9
745
EF653211.1
-
10
780
EF653212.1
-
9
674
EF653213.1
-
9
674
EF653214.1
-
10
780
EF653215.1
-
9
753
EF653216.1
-
8
639
EF653217.1
-
9
674
EF653218.1
-
9
674
EF653219.1
-
9
674
EF653220.1
-
9
674
EF653221.1
-
8
639
EF653222.1
-
10
754
EF653223.1
-
10
754
EF653224.1
-
9
674
EF685283.1
-
9
674
EF685284.1
-
9
674
EF685285.1
-
9
674
EF685286.1
-
9
674
EF685287.1
-
9
674
EF685288.1
-
9
674
EF685290.1
-
8
624
EF685291.1
-
10
788
EF685292.1
-
9
674
EF685293.1
-
10
788
EF685294.1
-
10
780
EF685295.1
-
9
674
EF685296.1
-
9
674
EF685297.1
-
9
674
EF685298.1
-
9
745
EF685299.1
-
10
780
EF685300.1
-
9
674
EF685301.1
-
9
674
EF685302.1
-
9
674
EF685303.1
-
9
674
EF685304.1
-
10
796
EF685305.1
-
10
749
EF685306.1
-
9
674
EF685307.1
-
11
894
EF685308.1
-
9
674
EF685309.1
-
9
674
EF685310.1
-
9
674
EF685311.1
-
9
674
EF685312.1
-
8
639
EF685313.1
-
8
639
EF685314.1
-
9
674
EF685316.1
-
9
674
EF685317.1
-
9
674
EF685318.1
-
9
674
EF685319.1
-
9
674
EF685320.1
-
9
674
EF685321.1
-
9
674
EF685322.1
-
9
674
EF685323.1
-
9
674
EF685324.1
-
9
674
EF685325.1
-
9
674
EF685326.1
-
9
674
EF685327.1
-
10
749
EF685328.1
-
9
674
EF685329.1
-
7
584
EF685330.1
-
9
674
EF685331.1
-
674
EF685332.1
-
9
674
EF685333.1
-
9
674
EF685334.1
-
9
674
EF685335.1
-
7
584
EF685336.1
-
9
674
EF685337.1
-
9
674
EF685338.1
-
9
674
EF685339.1
-
9
674
EF685340.1
-
9
674
EF685342.1
-
9
674
EU026275.1
-
10
780
EU026276.1
-
9
674
EU026277.1
-
9
674
EU026278.1
-
9
674
EU026279.1
-
7
584
EU026280.1
-
9
745
EU026281.1
-
8
639
EU026285.1
-
10
780
EU026286.1
-
8
639
EU026289.1
-
9
674
EU026290.1
-
7
584
EU026294.1
-
8
639
EU026295.1
-
7
584
EU026296.1
-
10
780
EU026297.1
-
10
780
EU026298.1
-
8
639
EU026299.1
-
8
639
EU026300.1
-
8
639
EU026301.1
-
9
745
EU026302.1
-
10
803
EU026303.1
-
9
674
EU026304.1
-
9
674
EU026305.1
-
9
674
EU026306.1
-
10
788
EU026307.1
-
9
674
EU026308.1
-
9
745
EU026309.1
-
9
674
EU026310.1
-
9
674
EU026311.1
-
9
674
EU026313.1
-
9
674
EU026314.1
-
10
780
EU026315.1
-
8
639
EU026316.1
-
10
754
EU026317.1
-
9
674
EU026318.1
-
7
589
EU026319.1
-
9
674
EU026320.1
-
10
780
EU026321.1
-
9
674
EU026322.1
-
9
674
EU026323.1
-
8
639
EU026324.1
-
8
639
EU026325.1
-
9
674
EU026326.1
-
10
780
EU026327.1
-
9
745
EU026328.1
-
8
639
EU026330.1
-
7
584
EU026331.1
-
9
674
EU026332.1
-
8
639
EU026333.1
-
9
744
EU026334.1
-
9
747
EU026335.1
-
9
674
EU026337.1
-
8
639
EU026338.1
-
9
674
EU026339.1
-
8
639
EU026340.1
-
10
859
EU026342.1
-
7
589
EU026343.1
-
10
796
EU026344.1
-
8
639
EU026345.1
-
9
674
EU026346.1
-
8
624
EU026347.1
-
8
639
EU026348.1
-
9
674
EU026349.1
-
9
674
EU026350.1
-
9
674
EU026351.1
-
9
674
EU116343.1
-
4
194
EU116345.1
-
9
674
EU116348.1
-
8
519
EU116349.1
-
10
867
EU116350.1
-
8
544
EU116351.1
-
3
138
EU116352.1
-
9
674
EU116353.1
-
9
674
EU163941.1
-
9
745
EU163942.1
-
9
674
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0