Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022949.2 | NP_075238.2 | 6 | 673 | ||
GenBank mRNA | BC168728.1 | - | 6 | 673 | 6918710 | |
FQ209993.1 | - | 3 | 420 | |||
FQ210501.1 | - | 3 | 420 | |||
FQ221537.1 | - | 3 | 420 | |||
FQ222022.1 | - | 3 | 420 | |||
FQ222656.1 | - | 3 | 420 | |||
FQ222698.1 | - | 3 | 420 | |||
FQ223044.1 | - | 3 | 421 | |||
FQ223231.1 | - | 3 | 420 | |||
FQ223484.1 | - | 3 | 420 | |||
FQ224076.1 | - | 3 | 420 | |||
FQ225475.1 | - | 3 | 420 | |||
FQ225874.1 | - | 3 | 420 | |||
FQ229197.1 | - | 3 | 420 | |||
FQ233060.1 | - | 3 | 420 | |||
FQ234143.1 | - | 3 | 420 | |||
X94242.1 | - | 5 | 655 |