Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133320.1 | NP_579854.1 | 3 | 163 | ||
XM_006246571.2 | XP_006246633.1 | 3 | 572 | |||
XM_006246572.2 | XP_006246634.1 | 3 | 565 | |||
XM_006246573.2 | XP_006246635.1 | 3 | 313 | |||
XM_006246574.2 | XP_006246636.1 | 3 | 198 | |||
XM_006246575.2 | XP_006246637.1 | 3 | 676 | |||
XM_006246576.2 | XP_006246638.1 | 3 | 566 | |||
XM_006246578.2 | XP_006246640.1 | 3 | 312 | |||
GenBank mRNA | AY008298.1 | - | 3 | 163 | ||
BC070909.1 | - | 3 | 253 | 7103650 |