Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 12, 13 | 1800 | ||
NM_001321185.1 | NP_001308114.1 | 12, 13 | 1753 | |||
NM_007267.7 | NP_009198.4 | 12, 13 | 1704 | |||
XM_005256995.1 | XP_005257052.1 | 12, 13 | 1829 | |||
XM_011524255.1 | XP_011522557.1 | 11, 12 | 1731 | |||
XM_011524256.1 | XP_011522558.1 | 11, 12 | 1605 | |||
XM_011524257.2 | XP_011522559.1 | 6, 7 | 849 | |||
XM_017024109.1 | XP_016879598.1 | 10, 11 | 1520 | |||
GenBank mRNA | AB002405.2 | - | 4, 5 | 512 | ||
AK021738.1 | - | 10 | 1741 | |||
AK074065.1 | - | 12, 13 | 1703 | |||
AK096076.1 | - | 12, 13 | 1736 | |||
AK310734.1 | - | 11, 12 | 2124 | |||
AY057379.1 | - | 11, 12 | 1543 | |||
AY099356.1 | - | 13, 14 | 1702 | |||
AY236496.1 | - | 11, 12 | 1669 | |||
BC007766.1 | - | 11, 12 | 1443 | |||
BC018346.1 | - | 5, 6 | 668 | |||
BC023597.2 | - | 12, 13 | 1684 | 4566166 | ||
BC035648.1 | - | 5, 6 | 724 | 5575949 |