Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153622.2 | NP_705886.1 | 3, 4 | 374 | ||
XM_006253941.2 | XP_006254003.1 | 6, 7 | 1027 | |||
XM_006253942.2 | XP_006254004.1 | 6, 7 | 1029 | |||
XM_006253944.2 | XP_006254006.1 | 6, 7 | 1016 | |||
XM_006253946.2 | XP_006254008.1 | 5, 6 | 903 | |||
XM_006253947.2 | XP_006254009.1 | 6, 7 | 1033 | |||
XM_006253948.2 | XP_006254010.1 | 1, 2 | 904 | |||
XM_006253949.2 | XP_006254011.1 | 4, 5 | 786 | |||
XM_008771623.1 | XP_008769845.1 | 7, 8 | 1104 | |||
XM_008771624.1 | XP_008769846.1 | 7, 8 | 1092 | |||
XM_008771625.1 | XP_008769847.1 | 5, 6 | 862 | |||
GenBank mRNA | AB025223.1 | - | 3, 4 | 398 | ||
AB025224.1 | - | 3, 4 | 398 | |||
BC078710.1 | - | 3, 4 | 292 | 7110091 |