Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFB2 SIRNA Assays ›
          Gene Symbol:
          Tgfb2
          Gene Name:
          transforming growth factor, beta 2
          Gene Aliases:
          TGF-B2
          Chromosome Location:
          Chr.13: 105039639 - 105141689 on Build Rnor_6.0
          UniGene ID:
          Rn.24539
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 194995

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tgfb2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031131.1 NP_112393.1 1 405
          XM_006250448.2 XP_006250510.1 1 1315
          GenBank mRNA AF135598.1 - 1 99
          AF153012.2 - 1 405
          AF153013.1 - 1 405
          AJ132718.1 - 1 113
          BC100663.1 - 1 577

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          skeletal system development
          cartilage condensation
          regulation of cell growth
          blood vessel development
          eye development
          response to hypoxia
          kidney development
          epithelial to mesenchymal transition
          hair follicle development
          blood vessel remodeling
          heart morphogenesis
          heart valve morphogenesis
          protein phosphorylation
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          axon guidance
          salivary gland morphogenesis
          heart development
          skeletal muscle tissue development
          female pregnancy
          aging
          cell death
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          glial cell migration
          response to radiation
          response to cold
          response to wounding
          cardioblast differentiation
          response to organonitrogen compound
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          negative regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell growth
          cell migration
          signal transduction by protein phosphorylation
          hemopoiesis
          extracellular matrix organization
          collagen fibril organization
          positive regulation of cell growth
          negative regulation of cell growth
          lung development
          positive regulation of cell migration
          neutrophil chemotaxis
          thyroid gland development
          hindbrain development
          pancreas development
          hair follicle morphogenesis
          activation of protein kinase activity
          response to estradiol
          response to retinoic acid
          response to progesterone
          positive regulation of stress-activated MAPK cascade
          regulation of transforming growth factor beta2 production
          response to vitamin D
          positive regulation of cell adhesion mediated by integrin
          response to cytokine
          response to laminar fluid shear stress
          wound healing
          regulation of cell proliferation
          dopamine biosynthetic process
          response to drug
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          response to estrogen
          cell-cell junction organization
          positive regulation of integrin biosynthetic process
          positive regulation of ossification
          positive regulation of cell cycle
          positive regulation of heart contraction
          somatic stem cell division
          cell development
          digestive tract development
          embryonic digestive tract development
          neuron fate commitment
          neuron development
          embryonic neurocranium morphogenesis
          inner ear development
          negative regulation of epithelial cell proliferation
          positive regulation of protein secretion
          negative regulation of immune response
          positive regulation of immune response
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of cell division
          positive regulation of catagen
          positive regulation of cardioblast differentiation
          cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          face morphogenesis
          frontal suture morphogenesis
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          negative regulation of cartilage development
          positive regulation of activation-induced cell death of T cells
          extrinsic apoptotic signaling pathway
          regulation of extracellular matrix organization
          regulation of complement-dependent cytotoxicity
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          beta-amyloid binding
          receptor signaling protein serine/threonine kinase activity
          receptor binding
          type II transforming growth factor beta receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          type III transforming growth factor beta receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0