Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166022.1 | NP_001159494.1 | 3 | 234 | ||
NM_013137.2 | NP_037269.2 | 3 | 321 | |||
GenBank mRNA | BC166604.1 | - | 3 | 321 | ||
CB613563.1 | - | 3 | 234 | |||
DN935615.1 | - | 3 | 352 |