Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADGRL2 SIRNA Assays ›
          Gene Symbol:
          ADGRL2
          Gene Name:
          adhesion G protein-coupled receptor L2
          Gene Aliases:
          CIRL2, CL2, LEC1, LPHH1, LPHN2
          Chromosome Location:
          Chr.1: 81306155 - 81992738 on Build GRCh38
          UniGene ID:
          Hs.24212
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 19871

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADGRL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001297704.1 NP_001284633.1 3, 4 262
          NM_001297705.1 NP_001284634.1 2, 3 297
          NM_001297706.1 NP_001284635.1 2, 3 297
          NM_012302.3 NP_036434.1 2, 3 297
          XM_005270666.4 XP_005270723.1 2, 3 1018
          XM_005270668.2 XP_005270725.1 2, 3 314
          XM_006710485.3 XP_006710548.1 2, 3 1019
          XM_006710488.2 XP_006710551.1 2, 3 313
          XM_006710489.3 XP_006710552.1 2, 3 204
          XM_017000782.1 XP_016856271.1 6, 7 887
          XM_017000783.1 XP_016856272.1 3, 4 298
          XM_017000784.1 XP_016856273.1 2, 3 203
          XM_017000785.1 XP_016856274.1 2, 3 313
          XM_017000786.1 XP_016856275.1 2, 3 317
          XM_017000787.1 XP_016856276.1 3, 4 240
          XM_017000788.1 XP_016856277.1 3, 4 240
          XM_017000789.1 XP_016856278.1 3, 4 239
          XM_017000790.1 XP_016856279.1 3, 4 239
          XM_017000791.1 XP_016856280.1 2, 3 318
          XM_017000792.1 XP_016856281.1 2, 3 238
          XM_017000793.1 XP_016856282.1 3, 4 241
          XM_017000794.1 XP_016856283.1 3, 4 240
          XM_017000795.1 XP_016856284.1 3, 4 239
          XM_017000796.1 XP_016856285.1 2, 3 278
          XM_017000797.1 XP_016856286.1 2, 3 223
          XM_017000798.1 XP_016856287.1 2, 3 285
          GenBank mRNA AF104266.1 - 2, 3 297
          AF104938.1 - 2, 3 297
          AF104939.1 - 2, 3 297
          AJ131581.1 - 3, 4 261
          AK123422.1 - 6, 7 789

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          latrotoxin receptor activity
          carbohydrate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0