Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019371.1 | NP_062244.1 | 4 | 1207 | ||
GenBank mRNA | U06713.1 | - | 4 | 1207 |