Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19924
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
5, 6
717
NM_001281772.2
NP_001268701.1
7, 8
916
NM_001281773.2
NP_001268702.1
6, 7
863
NM_001281774.2
NP_001268703.1
6, 7
863
NM_001281775.2
NP_001268704.1
5, 6
792
NM_001281776.2
NP_001268705.1
5, 6
792
NM_001281777.2
NP_001268706.1
5, 6
717
NM_001281778.2
NP_001268707.1
5, 6
717
NM_001281779.2
NP_001268708.1
5, 6
792
NM_001281780.2
NP_001268709.1
5, 6
792
NM_001281781.2
NP_001268710.1
5, 6
717
NM_001281782.2
NP_001268711.1
5, 6
717
NM_001281783.2
NP_001268712.1
5, 6
792
NM_001281784.2
NP_001268713.1
5, 6
717
NM_012408.5
NP_036540.3
5, 6
792
NM_183047.3
NP_898868.1
5, 6
792
NM_183048.3
NP_898869.1
5, 6
717
XM_005260356.4
XP_005260413.1
5, 6
829
XM_005260358.2
XP_005260415.1
4, 5
571
XM_005260360.4
XP_005260417.1
5, 6
1294
XM_005260362.2
XP_005260419.1
5, 6
569
XM_005260364.2
XP_005260421.1
5, 6
554
XM_005260366.2
XP_005260423.1
5, 6
585
XM_006723760.3
XP_006723823.1
5, 6
967
XM_006723762.3
XP_006723825.1
5, 6
668
XM_006723763.1
XP_006723826.1
5, 6
583
XM_006723766.1
XP_006723829.1
5, 6
527
XM_011528749.2
XP_011527051.1
5, 6
1473
XM_011528751.1
XP_011527053.1
5, 6
577
XM_011528752.1
XP_011527054.1
4, 5
559
XM_011528753.1
XP_011527055.1
6, 7
588
XM_011528754.1
XP_011527056.1
5, 6
515
XM_017027760.1
XP_016883249.1
4, 5
574
XM_017027761.1
XP_016883250.1
5, 6
1040
XM_017027762.1
XP_016883251.1
7, 8
1807
XM_017027763.1
XP_016883252.1
5, 6
720
XM_017027764.1
XP_016883253.1
5, 6
1239
XM_017027765.1
XP_016883254.1
5, 6
569
XM_017027766.1
XP_016883255.1
5, 6
569
XM_017027767.1
XP_016883256.1
5, 6
1384
XM_017027768.1
XP_016883257.1
5, 6
644
XM_017027769.1
XP_016883258.1
5, 6
554
XM_017027770.1
XP_016883259.1
6, 7
588
XM_017027771.1
XP_016883260.1
5, 6
554
XM_017027772.1
XP_016883261.1
5, 6
527
XM_017027773.1
XP_016883262.1
5, 6
569
XM_017027774.1
XP_016883263.1
4, 5
547
XM_017027775.1
XP_016883264.1
5, 6
515
XM_017027776.1
XP_016883265.1
5, 6
554
XM_017027777.1
XP_016883266.1
4, 5
547
XM_017027778.1
XP_016883267.1
4, 5
559
XM_017027779.1
XP_016883268.1
4, 5
559
XM_017027780.1
XP_016883269.1
4, 5
574
GenBank mRNA
AB032951.1
-
5, 6
767
AF454056.1
-
5, 6
609
AK000275.1
-
5, 6
534
AK225857.1
-
5, 6
600
AK294511.1
-
7, 8
726
AK295747.1
-
5, 6
676
AK299899.1
-
6, 7
675
AK309228.1
-
5, 6
586
AL137703.1
-
4, 5
529
BC030721.1
-
2, 3
292
BC136608.1
-
5, 6
548
9020598
BC144289.1
-
5, 6
548
9052809
BC146802.1
-
5, 6
558
8860316
BI553958.1
-
5, 6
563
BX641005.1
-
6, 7
808
DQ082998.1
-
4, 5
573
DQ082999.1
-
4, 5
573
DQ368669.1
-
4, 5
573
DQ368670.1
-
4, 5
573
DQ368671.1
-
4, 5
498
DQ368672.1
-
4, 5
498
DQ368673.1
-
4, 5
573
DQ368674.1
-
4, 5
498
DQ368675.1
-
4, 5
573
DQ368676.1
-
4, 5
573
DQ368677.1
-
4, 5
498
DQ368678.1
-
4, 5
498
DQ368679.1
-
4, 5
573
DQ368680.1
-
4, 5
498
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0