Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012608.2 | NP_036740.1 | 9 | 866 | ||
XM_006232437.2 | XP_006232499.1 | 9 | 902 | |||
GenBank mRNA | BC085753.1 | - | 9 | 866 | 7110842 | |
M15944.1 | - | 8 | 816 |