Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012727.3 | NP_036859.2 | 3 | 352 | ||
XM_006254523.2 | XP_006254585.1 | 2 | 294 | |||
GenBank mRNA | BC128706.1 | - | 3 | 352 | 7455982 | |
M63334.1 | - | 246 | ||||
M64757.1 | - | 1 | 342 | |||
S65840.1 | - | 2 | 277 |