Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012765.3 | NP_036897.2 | 4 | 445 | ||
XM_008773402.1 | XP_008771624.1 | 4 | 814 | |||
GenBank mRNA | U35315.1 | - | 4 | 444 |
Set Membership: |
![]() |