Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012788.1 | NP_036920.1 | 3 | 591 | ||
GenBank mRNA | U14950.1 | - | 3 | 591 |