Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP2CA SIRNA Assays ›
          Gene Symbol:
          Ppp2ca LOC103694903
          Gene Name:
          protein phosphatase 2, catalytic subunit, alpha isozyme
          serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
          Gene Aliases:
          Pp2a1
          -
          Chromosome Location:
          Chr.10: 37535108 - 37554861 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906910: 112829 - 133730 on Build Rnor_6.0
          UniGene ID:
          Rn.1271
          Rn.1271
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 200660

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ppp2ca
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017039.2 NP_058735.1 3 609
          GenBank mRNA BC070914.1 - 3 618 7104697
          BC072531.1 - 3 609 7096821
          FQ228785.1 - 2 173
          M33114.1 - 3 525
          X14159.1 - 3 525
          X16043.1 - 3 584
          Gene Symbol: LOC103694903
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773894.1 XP_008772116.1 1104

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_019411, AK172644, BC003856, AK076110, AF076192, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of protein phosphorylation
          negative regulation of protein phosphorylation
          cardiac ventricle development
          protein dephosphorylation
          mesoderm development
          heart development
          regulation of receptor activity
          negative regulation of epithelial to mesenchymal transition
          regulation of protein autophosphorylation
          cellular response to insulin stimulus
          positive regulation of protein phosphatase type 2A activity
          positive regulation of protein dephosphorylation
          regulation of protein catabolic process
          negative regulation of transcription factor import into nucleus
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          meiotic cell cycle
          protein heterotrimerization
          cellular response to calcium ion
          cellular response to glucose stimulus
          cellular response to cytokine stimulus
          cellular response to ethanol
          cellular response to follicle-stimulating hormone stimulus
          cellular response to steroid hormone stimulus
          positive regulation of protein serine/threonine kinase activity
          negative regulation of calcium ion transmembrane transporter activity
          protein serine/threonine phosphatase activity
          protein binding
          protein C-terminus binding
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          protein complex binding
          protein kinase B binding
          ion channel binding
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity
          tau protein binding
          GABA receptor binding
          protein phosphatase 2A binding
          protein antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix