Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT5A SIRNA Assays ›
          Gene Symbol:
          Stat5a
          Gene Name:
          signal transducer and activator of transcription 5A
          Gene Aliases:
          Stat5
          Chromosome Location:
          Chr.10: 88764732 - 88789060 on Build Rnor_6.0
          UniGene ID:
          Rn.154399
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200722

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Stat5a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017064.1 NP_058760.1 16 1990
          GenBank mRNA U24175.1 - 16 1990

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin metabolic process
          luteinization
          natural killer cell differentiation
          citrate metabolic process
          2-oxoglutarate metabolic process
          succinate metabolic process
          oxaloacetate metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          isoleucine metabolic process
          valine metabolic process
          creatine metabolic process
          fatty acid metabolic process
          signal transduction
          JAK-STAT cascade
          female pregnancy
          lactation
          positive regulation of cell proliferation
          peptidyl-tyrosine phosphorylation
          regulation of steroid metabolic process
          cytokine-mediated signaling pathway
          taurine metabolic process
          lipid storage
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          mammary gland development
          response to nutrient levels
          positive regulation of natural killer cell differentiation
          negative regulation of mast cell apoptotic process
          T cell differentiation in thymus
          prolactin signaling pathway
          regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          T cell homeostasis
          negative regulation of apoptotic process
          response to peptide hormone
          positive regulation of interleukin-2 biosynthetic process
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of lymphocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of natural killer cell mediated cytotoxicity
          creatinine metabolic process
          development of secondary female sexual characteristics
          development of secondary male sexual characteristics
          Peyer's patch development
          multicellular organismal reproductive process
          positive regulation of inflammatory response
          mammary gland involution
          positive regulation of mast cell differentiation
          JAK-STAT cascade involved in growth hormone signaling pathway
          prostate gland epithelium morphogenesis
          epithelial cell differentiation involved in prostate gland development
          mammary gland epithelium development
          positive regulation of mast cell proliferation
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          protein binding
          protein phosphatase binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0