Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200849
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
3
361
NM_001034069.1
NP_001029241.1
3
361
NM_001034070.1
NP_001029242.1
3
361
NM_001034071.1
NP_001029243.1
3
361
NM_001034072.1
NP_001029244.1
3
361
NM_001034073.1
NP_001029245.1
2
298
NM_001034074.1
NP_001029246.1
2
298
NM_001034075.1
NP_001029247.1
2
298
NM_001301336.1
NP_001288265.1
3
361
NM_001301342.1
NP_001288271.1
3
361
NM_001301736.1
NP_001288665.1
2
298
NM_019131.2
NP_062004.1
2
298
XM_006243386.2
XP_006243448.1
3
353
XM_006243388.2
XP_006243450.1
3
353
XM_006243389.2
XP_006243451.1
3
353
XM_006243390.2
XP_006243452.1
3
353
XM_006243393.1
XP_006243455.1
2
296
XM_006243394.1
2
314
XM_006243395.2
XP_006243457.1
2
305
XM_008766207.1
XP_008764429.1
3
353
XM_008766208.1
XP_008764430.1
2
296
GenBank mRNA
AF372215.1
-
2
147
AF372216.1
-
3
260
AF372217.1
-
3
266
BC078780.1
-
2
265
7106337
BU670983.1
-
3
361
CB585296.1
-
2
301
CB787774.1
-
3
407
CF108680.1
-
3
309
CK483405.1
-
3
315
CO562261.1
-
328
FQ074026.1
-
3
419
FQ214619.1
-
3
336
FQ214803.1
-
3
332
FQ214809.1
-
3
330
FQ214835.1
-
3
330
FQ214864.1
-
3
334
FQ214868.1
-
3
330
FQ214869.1
-
3
336
FQ214872.1
-
3
329
FQ214877.1
-
3
330
FQ214894.1
-
3
336
FQ214902.1
-
3
330
FQ214911.1
-
3
332
FQ214944.1
-
3
336
FQ214946.1
-
3
336
FQ214991.1
-
3
330
FQ215107.1
-
3
330
FQ215186.1
-
3
336
FQ215188.1
-
3
330
FQ215206.1
-
3
365
FQ215226.1
-
3
336
FQ215459.1
-
3
332
FQ215512.1
-
3
336
FQ215522.1
-
3
407
FQ215524.1
-
3
332
FQ215538.1
-
3
336
FQ215551.1
-
3
330
FQ215698.1
-
3
330
FQ215703.1
-
3
330
FQ215724.1
-
3
417
FQ215746.1
-
3
332
FQ215761.1
-
3
330
FQ215803.1
-
3
336
FQ215808.1
-
3
332
FQ215815.1
-
3
330
FQ215856.1
-
3
330
FQ215919.1
-
3
338
FQ215944.1
-
3
336
FQ215969.1
-
3
330
FQ215995.1
-
3
332
FQ216014.1
-
3
414
FQ216018.1
-
3
336
FQ216052.1
-
3
330
FQ216105.1
-
3
330
FQ216118.1
-
3
414
FQ216145.1
-
3
336
FQ216160.1
-
3
329
FQ216319.1
-
3
332
FQ216340.1
-
3
336
FQ216368.1
-
3
332
FQ216400.1
-
3
332
FQ216494.1
-
3
330
FQ216552.1
-
3
336
FQ216557.1
-
3
336
FQ216569.1
-
424
FQ216615.1
-
3
245
FQ216637.1
-
3
332
FQ216683.1
-
3
332
FQ216697.1
-
3
332
FQ216747.1
-
3
332
FQ217468.1
-
2
81
FQ217503.1
-
3
336
FQ217637.1
-
3
336
FQ217808.1
-
3
332
FQ223706.1
-
3
332
FQ223993.1
-
3
334
FQ224036.1
-
3
247
FQ224207.1
-
3
330
FQ224838.1
-
3
336
FQ224943.1
-
3
334
FQ224961.1
-
3
332
M34134.1
-
2
283
M34135.1
-
3
295
M34136.1
-
1
13
M60666.1
-
3
308
M60667.1
-
3
308
M60668.1
-
2
245
M60669.1
-
2
245
X02411.1
-
3
259
X02412.1
-
1
15
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0