Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200851
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
3
366
NM_001034069.1
NP_001029241.1
3
366
NM_001034070.1
NP_001029242.1
3
366
NM_001034071.1
NP_001029243.1
3
366
NM_001034072.1
NP_001029244.1
3
366
NM_001034073.1
NP_001029245.1
2
303
NM_001034074.1
NP_001029246.1
2
303
NM_001034075.1
NP_001029247.1
2
303
NM_001301336.1
NP_001288265.1
3
366
NM_001301342.1
NP_001288271.1
3
366
NM_001301736.1
NP_001288665.1
2
303
NM_019131.2
NP_062004.1
2
303
XM_006243386.2
XP_006243448.1
3
358
XM_006243388.2
XP_006243450.1
3
358
XM_006243389.2
XP_006243451.1
3
358
XM_006243390.2
XP_006243452.1
3
358
XM_006243393.1
XP_006243455.1
2
301
XM_006243394.1
2
319
XM_006243395.2
XP_006243457.1
2
310
XM_008766207.1
XP_008764429.1
3
358
XM_008766208.1
XP_008764430.1
2
301
GenBank mRNA
AF372215.1
-
2
152
AF372216.1
-
3
265
AF372217.1
-
3
271
BC078780.1
-
2
270
7106337
BU670983.1
-
3
366
CB585296.1
-
2
306
CB787774.1
-
3
412
CF108680.1
-
3
314
CK483405.1
-
3
320
CO562261.1
-
333
FQ074026.1
-
3
424
FQ214619.1
-
3
341
FQ214803.1
-
3
337
FQ214809.1
-
3
335
FQ214835.1
-
3
335
FQ214864.1
-
3
339
FQ214868.1
-
3
335
FQ214869.1
-
3
341
FQ214872.1
-
3
334
FQ214877.1
-
3
335
FQ214894.1
-
3
341
FQ214902.1
-
3
335
FQ214911.1
-
3
337
FQ214944.1
-
3
341
FQ214946.1
-
3
341
FQ214991.1
-
3
335
FQ215107.1
-
3
335
FQ215186.1
-
3
341
FQ215188.1
-
3
335
FQ215206.1
-
3
370
FQ215226.1
-
3
341
FQ215459.1
-
3
337
FQ215512.1
-
3
341
FQ215522.1
-
3
412
FQ215524.1
-
3
337
FQ215538.1
-
3
341
FQ215551.1
-
3
335
FQ215698.1
-
3
335
FQ215703.1
-
3
335
FQ215724.1
-
3
422
FQ215746.1
-
3
337
FQ215761.1
-
3
335
FQ215768.1
-
3
335
FQ215803.1
-
3
341
FQ215808.1
-
3
337
FQ215815.1
-
3
335
FQ215856.1
-
3
335
FQ215919.1
-
3
343
FQ215944.1
-
3
341
FQ215969.1
-
3
335
FQ215995.1
-
3
337
FQ216014.1
-
3
419
FQ216018.1
-
3
341
FQ216052.1
-
3
335
FQ216082.1
-
3
419
FQ216105.1
-
3
335
FQ216118.1
-
3
419
FQ216145.1
-
3
341
FQ216160.1
-
3
334
FQ216319.1
-
3
337
FQ216340.1
-
3
341
FQ216368.1
-
3
337
FQ216400.1
-
3
337
FQ216494.1
-
3
335
FQ216552.1
-
3
341
FQ216557.1
-
3
341
FQ216569.1
-
429
FQ216615.1
-
3
250
FQ216637.1
-
3
337
FQ216683.1
-
3
337
FQ216697.1
-
3
337
FQ216747.1
-
3
337
FQ217468.1
-
2
86
FQ217503.1
-
3
341
FQ217637.1
-
3
341
FQ217808.1
-
3
337
FQ223706.1
-
3
337
FQ223993.1
-
3
339
FQ224036.1
-
3
252
FQ224207.1
-
3
335
FQ224838.1
-
3
341
FQ224943.1
-
3
339
FQ224961.1
-
3
337
M34134.1
-
2
288
M34135.1
-
3
300
M34136.1
-
1
18
M60666.1
-
3
313
M60667.1
-
3
313
M60668.1
-
2
250
M60669.1
-
2
250
X02411.1
-
3
264
X02412.1
-
1
20
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0