Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024127.2 | NP_077041.2 | 4 | 697 | ||
GenBank mRNA | BC081795.1 | - | 4 | 697 | 7110642 | |
L32591.1 | - | 4 | 565 |