Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOS1 SIRNA Assays ›
          Gene Symbol:
          Nos1
          Gene Name:
          nitric oxide synthase 1, neuronal
          Gene Aliases:
          bNOS
          Chromosome Location:
          Chr.12: 44215395 - 44405539 on Build Rnor_6.0
          UniGene ID:
          Rn.10573
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 201230

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nos1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_052799.1 NP_434686.1 1 223
          XM_008769210.1 XP_008767432.1 2 1034
          GenBank mRNA U67309.1 - 1 223
          X59949.1 - 1 470

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of sodium ion transport
          arginine catabolic process
          nitric oxide biosynthetic process
          muscle contraction
          striated muscle contraction
          nitric oxide mediated signal transduction
          female pregnancy
          aging
          regulation of heart contraction
          negative regulation of cell proliferation
          response to heat
          response to organonitrogen compound
          response to lead ion
          response to organic cyclic compound
          response to activity
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to nutrient levels
          response to lipopolysaccharide
          negative regulation of peptidyl-serine phosphorylation
          response to vitamin E
          multicellular organismal response to stress
          positive regulation of histone acetylation
          response to nicotine
          exogenous drug catabolic process
          negative regulation of apoptotic process
          negative regulation of potassium ion transport
          response to peptide hormone
          response to estrogen
          response to ethanol
          negative regulation of blood pressure
          negative regulation of heart contraction
          positive regulation of transcription, DNA-templated
          negative regulation of vasoconstriction
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          behavioral response to cocaine
          negative regulation of hydrolase activity
          negative regulation of cytosolic calcium ion concentration
          negative regulation of serotonin uptake
          negative regulation of calcium ion transport
          regulation of sensory perception of pain
          oxidation-reduction process
          cellular response to mechanical stimulus
          cellular response to growth factor stimulus
          response to nitric oxide
          cellular response to epinephrine stimulus
          positive regulation of the force of heart contraction
          positive regulation of long-term synaptic potentiation
          positive regulation of adrenergic receptor signaling pathway involved in heart process
          positive regulation of neuron death
          positive regulation of sodium ion transmembrane transport
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          iron ion binding
          protein binding
          calmodulin binding
          FMN binding
          amino acid binding
          sodium channel regulator activity
          enzyme binding
          heme binding
          protein homodimerization activity
          ion channel binding
          cadmium ion binding
          flavin adenine dinucleotide binding
          NADP binding
          ATPase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0