Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130399.2 | NP_569083.1 | 5 | 511 | ||
GenBank mRNA | AB059655.1 | - | 5 | 435 | ||
BC088116.1 | - | 5 | 511 | 7384286 |