Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139263.1 | NP_640356.1 | 9 | 2446 | ||
XM_008768850.1 | XP_008767072.1 | 9 | 1822 | |||
GenBank mRNA | AF038388.1 | - | 9 | 2446 |