Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 8 | 1182 | ||
XM_011536675.2 | XP_011534977.1 | 8 | 1182 | |||
XM_011536676.2 | XP_011534978.1 | 7 | 1095 | |||
XM_011536679.2 | XP_011534981.1 | 4 | 384 | |||
XM_017021244.1 | XP_016876733.1 | 7 | 1016 | |||
XM_017021245.1 | XP_016876734.1 | 4 | 470 | |||
GenBank mRNA | AB209114.1 | - | 8 | 1004 | ||
AF105020.1 | - | 8 | 1185 | |||
AK023804.1 | - | 2 | 1773 | |||
AY090480.1 | - | 8 | 1185 | |||
BC031651.1 | - | 8 | 1072 | 5169145 | ||
BM127609.1 | - | 2 | 106 | |||
CR627447.1 | - | 8 | 752 |