Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243270.1 | NP_001230199.1 | 4 | 727 | ||
NM_001243271.1 | NP_001230200.1 | 5 | 866 | |||
NM_014361.3 | NP_055176.1 | 5 | 866 | |||
NM_175566.2 | NP_780775.1 | 2 | 210 | |||
XM_011542871.1 | XP_011541173.1 | 4 | 644 | |||
XM_011542873.1 | XP_011541175.1 | 5 | 866 | |||
XM_017017926.1 | XP_016873415.1 | 6 | 1034 | |||
XM_017017927.1 | XP_016873416.1 | 6 | 1034 | |||
XM_017017928.1 | XP_016873417.1 | 4 | 799 | |||
XM_017017929.1 | XP_016873418.1 | 4 | 644 | |||
GenBank mRNA | AB013802.2 | - | 3 | 441 | ||
AB013803.2 | - | 2 | 219 | |||
AK308788.1 | - | 5 | 866 | |||
BC039255.1 | - | 5 | 701 | 5273769 | ||
BC130619.1 | - | 4 | 513 | 40146650 | ||
BC144185.1 | - | 4 | 513 | 9052705 | ||
BC144186.1 | - | 5 | 652 | 9052706 |