Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014641.2 | NP_055456.2 | 3 | 795 | ||
XM_005249492.2 | XP_005249549.1 | 3 | 699 | |||
XM_005249493.3 | XP_005249550.1 | 3 | 699 | |||
XM_005249494.4 | XP_005249551.1 | 3 | 1098 | |||
XM_005249497.3 | XP_005249554.1 | 3 | 699 | |||
XM_005249498.3 | XP_005249555.1 | 3 | 699 | |||
XM_011515001.2 | XP_011513303.1 | 3 | 526 | |||
XM_011515003.2 | XP_011513305.1 | 3 | 381 | |||
XM_017011519.1 | XP_016867008.1 | 3 | 478 | |||
XM_017011520.1 | XP_016867009.1 | 3 | 397 | |||
XM_017011521.1 | XP_016867010.1 | 3 | 397 | |||
XM_017011522.1 | XP_016867011.1 | 3 | 699 | |||
GenBank mRNA | BC131491.1 | - | 3 | 795 | ||
BC152556.1 | - | 2 | 368 | 9007258 | ||
CR749828.1 | - | 3 | 473 | |||
D79992.1 | - | 2 | 368 | |||
EF177820.1 | - | 3 | 423 | |||
EF177821.1 | - | 3 | 497 |