Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAV2 SIRNA Assays ›
Gene Symbol:
NAV2
Gene Name:
neuron navigator 2
Gene Aliases:
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location:
Chr.11: 19345200 - 20121601 on Build GRCh38
UniGene ID:
Hs.502116
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215794
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAV2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001111018.1
NP_001104488.1
20
4907
NM_001111019.2
NP_001104489.1
10
2319
NM_001244963.1
NP_001231892.1
22
5399
NM_145117.4
NP_660093.2
20
5231
NM_182964.5
NP_892009.3
20
5231
XM_005253214.4
XP_005253271.1
8
6706
XM_006718364.3
XP_006718427.1
21
5652
XM_006718365.3
XP_006718428.1
21
5622
XM_006718366.3
XP_006718429.1
22
4885
XM_006718367.3
XP_006718430.1
11
6919
XM_006718368.3
XP_006718431.1
9
6751
XM_006718369.2
XP_006718432.1
11
2414
XM_011520444.2
XP_011518746.1
22
5721
XM_011520445.2
XP_011518747.1
21
5655
XM_011520446.2
XP_011518748.1
21
5622
XM_011520447.2
XP_011518749.1
20
5610
XM_011520448.2
XP_011518750.1
22
5592
XM_011520449.2
XP_011518751.1
20
5556
XM_011520450.2
XP_011518752.1
19
5511
XM_011520451.2
XP_011518753.1
21
5099
XM_011520452.2
XP_011518754.1
21
5370
XM_011520453.2
XP_011518755.1
11
2349
XM_011520454.2
XP_011518756.1
8
2139
XM_017018520.1
XP_016874009.1
23
5513
XM_017018521.1
XP_016874010.1
19
5511
XM_017018522.1
XP_016874011.1
22
5469
XM_017018523.1
XP_016874012.1
22
6398
XM_017018524.1
XP_016874013.1
22
5221
XM_017018525.1
XP_016874014.1
16
4064
XM_017018526.1
XP_016874015.1
12
4700
GenBank mRNA
AB037840.3
-
20
5232
AB063115.1
-
20
4907
AB063116.1
-
20
5200
AF466143.1
-
20
4946
AF466144.1
-
20
4946
AJ488102.1
-
22
5137
AK074287.1
-
10
2319
AK290458.1
-
4
521
AK298346.1
-
8
2286
AK307574.1
-
8
2102
BC016054.1
-
2
227
BC150290.1
-
20
5232
8860130
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by baroreceptor feedback
sensory perception of sound
sensory perception of smell
locomotory behavior
optic nerve development
glossopharyngeal nerve development
vagus nerve development
helicase activity
protein binding
ATP binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0