Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308155.1 | NP_001295084.1 | 3, 4 | 959 | ||
NM_014498.4 | NP_055313.1 | 3, 4 | 959 | |||
XM_005247364.2 | XP_005247421.1 | 3, 4 | 958 | |||
XM_005247365.2 | XP_005247422.1 | 3, 4 | 958 | |||
XM_017006183.1 | XP_016861672.1 | 3, 4 | 438 | |||
GenBank mRNA | AK307557.1 | - | 3, 4 | 664 | ||
AK307718.1 | - | 3, 4 | 634 | |||
U55853.1 | - | 3, 4 | 332 |