Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 68 | 17462 | ||
XM_005257545.4 | XP_005257602.2 | 69 | 17650 | |||
XM_005257546.4 | XP_005257603.2 | 69 | 17554 | |||
XM_006721995.3 | XP_006722058.1 | 69 | 17937 | |||
XM_017024905.1 | XP_016880394.1 | 64 | 16517 | |||
GenBank mRNA | AB046774.1 | - | 28 | 5652 | ||
AB537889.1 | - | 68 | 17455 | |||
AK023871.1 | - | 11 | 3281 | |||
AK026038.1 | - | 7 | 2648 | |||
AL161961.1 | - | 6 | 3121 | |||
AL833201.1 | - | 28 | 5679 | |||
BC032220.1 | - | 12 | 3425 | |||
BX640932.1 | - | 34 | 6618 | |||
BX647946.1 | - | 40 | 8253 |