Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
217077
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
11
2000
NM_001042358.4
NP_001035817.1
10
1858
NM_001042359.3
NP_001035818.1
8
1432
NM_001042360.3
NP_001035819.1
5
1309
NM_001042361.4
NP_001035820.1
8
1307
NM_001042362.3
NP_001035821.1
6
1064
NM_001042363.4
NP_001035822.1
10
1843
NM_001042364.4
NP_001035823.1
7
1197
NM_001042365.3
NP_001035824.1
4
856
NM_001320681.1
NP_001307610.1
9
1748
NM_001320682.1
NP_001307611.1
5
954
NM_001320683.1
NP_001307612.1
9
1464
NM_001320684.1
NP_001307613.1
8
1373
NM_001320685.1
NP_001307614.1
10
1920
NM_001320686.1
NP_001307615.1
9
1763
NM_001320688.1
NP_001307617.1
7
1337
NM_001320689.1
NP_001307618.1
6
1105
NM_001320690.1
NP_001307619.1
5
969
NM_001320691.1
NP_001307620.1
8
1672
NM_015605.8
NP_056420.3
9
1449
XM_011539605.2
XP_011537907.1
11
2144
XM_011539606.2
XP_011537908.1
11
3236
XM_011539607.2
XP_011537909.1
11
2135
XM_011539610.2
XP_011537912.1
8
1618
XM_011539611.2
XP_011537913.1
7
1618
XM_017016042.1
XP_016871531.1
11
2081
XM_017016044.1
XP_016871533.1
10
1901
XM_017016045.1
XP_016871534.1
5
3220
XM_017016046.1
XP_016871535.1
10
1763
XM_017016049.1
XP_016871538.1
8
1494
XM_017016050.1
XP_016871539.1
6
1361
XM_017016053.1
XP_016871542.1
8
1337
XM_017016054.1
XP_016871543.1
8
1350
XM_017016055.1
XP_016871544.1
8
1439
XM_017016056.1
XP_016871545.1
7
1345
XM_017016057.1
XP_016871546.1
8
1655
XM_017016058.1
XP_016871547.1
6
1381
XM_017016059.1
XP_016871548.1
7
1339
XM_017016060.1
XP_016871549.1
7
1185
XM_017016061.1
XP_016871550.1
6
1135
XM_017016062.1
XP_016871551.1
6
1255
XM_017016063.1
XP_016871552.1
7
1226
XM_017016064.1
XP_016871553.1
5
978
XM_017016065.1
XP_016871554.1
6
1028
GenBank mRNA
AL050040.1
-
9
1706
BX648913.1
-
4
857
CR933668.1
-
5
922
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0