Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP2 SIRNA Assays ›
Gene Symbol:
KCNIP2
Gene Name:
potassium voltage-gated channel interacting protein 2
Gene Aliases:
KCHIP2
Chromosome Location:
Chr.10: 101825971 - 101843920 on Build GRCh38
UniGene ID:
Hs.97044
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21711
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014591.4
NP_055406.2
9
1146
NM_173191.2
NP_775283.1
9
1101
NM_173192.2
NP_775284.1
8
1047
NM_173193.2
NP_775285.1
7
972
NM_173194.2
NP_775286.1
7
628
NM_173195.2
NP_775287.1
7
951
NM_173197.2
NP_775289.1
6
843
XM_005269729.2
XP_005269786.1
8
949
XM_005269730.2
XP_005269787.1
7
906
XM_006717812.2
XP_006717875.1
9
1003
XM_011539731.2
XP_011538033.1
7
643
XM_017016161.1
XP_016871650.1
7
698
GenBank mRNA
AB044584.1
-
9
749
AB044585.1
-
7
599
AF199598.1
-
8
695
AF295076.1
-
7
599
AF295530.1
-
7
620
AF347114.1
-
7
599
AF367018.1
-
9
794
AF367019.1
-
6
491
AF367020.1
-
5
470
AF367021.1
-
9
749
AJ276317.1
-
2
114
AK027347.1
-
7
619
AK055089.1
-
9
1025
AK094668.1
-
11
1153
AK125382.1
-
7
1658
AK295324.1
-
5
582
AK301433.1
-
7
620
AK309368.1
-
7
827
AK314536.1
-
7
832
AK315042.1
-
8
927
AL136722.1
-
7
646
AY026328.1
-
7
599
AY302141.1
-
7
587
BC034685.1
-
8
910
4155871
BG911444.1
-
5
565
DQ148480.1
-
9
749
DQ148481.1
-
8
695
DQ148482.1
-
7
596
DQ148483.1
-
7
614
DQ148484.1
-
6
541
DQ862463.1
-
7
599
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145704, NM_001276358, NM_145703, NM_030716, AY647241, and others.
Gene Ontology Categories:
Function(s)
Process(es)
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix