Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANO4 SIRNA Assays ›
          Gene Symbol:
          ANO4
          Gene Name:
          anoctamin 4
          Gene Aliases:
          TMEM16D
          Chromosome Location:
          Chr.12: 100717457 - 101128641 on Build GRCh38
          UniGene ID:
          Hs.58785
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217322

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANO4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286615.1 NP_001273544.1 26, 27 3256
          NM_001286616.1 NP_001273545.1 25, 26 2824
          NM_178826.3 NP_849148.2 25, 26 3151
          XM_011537911.2 XP_011536213.1 28, 29 3251
          XM_011537912.2 XP_011536214.1 28, 29 3251
          XM_011537913.2 XP_011536215.1 27, 28 3206
          XM_011537914.2 XP_011536216.1 25, 26 2762
          XM_011537915.2 XP_011536217.1 26, 27 3074
          XM_011537916.2 XP_011536218.1 26, 27 2915
          XM_017018808.1 XP_016874297.1 25, 26 2762
          XM_017018809.1 XP_016874298.1 27, 28 3342
          XM_017018810.1 XP_016874299.1 25, 26 3004
          XM_017018811.1 XP_016874300.1 24, 25 2959
          XM_017018812.1 XP_016874301.1 24, 25 4977
          XM_017018813.1 XP_016874302.1 23, 24 2827
          XM_017018814.1 XP_016874303.1 26, 27 5216
          XM_017018815.1 XP_016874304.1 26, 27 5216
          XM_017018816.1 XP_016874305.1 26, 27 3299
          XM_017018817.1 XP_016874306.1 26, 27 5251
          GenBank mRNA AK091540.1 - 16, 17 1772
          AK091591.1 - 25, 26 2940
          AK092596.1 - 13, 14 1338
          AK293810.1 - 26, 27 2894
          AK316105.1 - 26, 27 2895
          BC109308.2 - 25, 26 2809 40035759

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106778, XM_008765230, XM_008765227, XM_008765228, XM_008765229, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001277188, BC065114.

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          lipid transport
          ion transmembrane transport
          calcium activated phosphatidylserine scrambling
          calcium activated phosphatidylcholine scrambling
          calcium activated galactosylceramide scrambling
          chloride transmembrane transport
          intracellular calcium activated chloride channel activity
          phospholipid scramblase activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix